5-Bromo-2,3-difluorophenylacetic acid 95%
Lieferant: Apollo Scientific
PC303426-1G
PC303426-5G
APOSPC303426-1GEA
451
EUR
APOSPC303426-1G
APOSPC303426-5G
5-Bromo-2,3-difluorophenylacetic acid 95%
5-Bromo-2,3-difluorophenylacetic acid
Formel:
C₈H₅BrF₂O₂ Molecular Weight: 251,025 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28131371 CAS-Nummer: 1780302-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...