5-Bromo-2,3-difluorobenzoyl chloride

Lieferant: Apollo Scientific

PC303429-1G PC303429-5G
APOSPC303429-1GEA 209 EUR
APOSPC303429-1G APOSPC303429-5G
5-Bromo-2,3-difluorobenzoyl chloride
5-Bromo-2,3-difluorobenzoyl chloride
Formel: C₇H₂BrClF₂O
Molecular Weight: 255,444 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737912
CAS-Nummer: 1807172-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR