6-Bromo-2,3-difluorobenzyl alcohol

Lieferant: Apollo Scientific

Synonyme: (6-Bromo-2,3-difluorophenyl)methanol

PC303432-1G PC303432-5G
APOSPC303432-1GEA 63.8 EUR
APOSPC303432-1G APOSPC303432-5G
6-Bromo-2,3-difluorobenzyl alcohol
6-Bromo-2,3-difluorobenzyl alcohol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368109
CAS-Nummer: 651326-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR