6-Bromo-2,3-difluorophenylacetic acid

Lieferant: Apollo Scientific

Synonyme: 2-(6-Bromo-2,3-difluorophenyl)acetic acid

PC303434-1G
APOSPC303434-1GEA 432 EUR
APOSPC303434-1G
6-Bromo-2,3-difluorophenylacetic acid
6-Bromo-2,3-difluorophenylacetic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368100
CAS-Nummer: 887585-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR