5-Bromo-3-chloro-2-fluorobenzyl alcohol

Lieferant: Apollo Scientific

Synonyme: (5-Bromo-3-chloro-2-fluorophenyl)methanol

PC303453-1G PC303453-5G
APOSPC303453-1GEA 231 EUR
APOSPC303453-1G APOSPC303453-5G
5-Bromo-3-chloro-2-fluorobenzyl alcohol
5-Bromo-3-chloro-2-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239.469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29036608
CAS-Nummer: 1514304-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR