3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific

PC303466-1G PC303466-5G PC303466-25G
APOSPC303466-1GEA 279 EUR
APOSPC303466-1G APOSPC303466-5G APOSPC303466-25G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₄Br₂F₄
Molecular Weight: 335.919 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32265218
CAS-Nummer: 2092564-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR