3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetic acid 97%
Lieferant: Apollo Scientific
PC303468-1G
PC303468-250MG
APOSPC303468-1GEA
522
EUR
APOSPC303468-1G
APOSPC303468-250MG
3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetic acid 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetic acid
Formel:
C₉H₅BrF₄O₂ Molecular Weight: 301.032 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32265229 CAS-Nummer: 2385623-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...