5-Bromo-4-chloro-2-fluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303479-1G PC303479-5G
APOSPC303479-1GEA 325 EUR
APOSPC303479-1G APOSPC303479-5G
5-Bromo-4-chloro-2-fluorophenylacetonitrile 97%
5-Bromo-4-chloro-2-fluorophenylacetonitrile
Formel: C₈H₄BrClFN
Molecular Weight: 248,479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28060843
CAS-Nummer: 1785359-01-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR