5-Bromo-4-chloro-2-fluorobenzamide
Lieferant: Apollo Scientific
PC303483-1G
PC303483-5G
APOSPC303483-1GEA
279
EUR
APOSPC303483-1G
APOSPC303483-5G
5-Bromo-4-chloro-2-fluorobenzamide
5-Bromo-4-chloro-2-fluorobenzamide
Formel:
C₇H₄BrClFNO Molecular Weight: 252.468 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28060774 CAS-Nummer: 1532518-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...