3-Bromo-5,6-difluoro-2-methoxybenzyl alcohol
Lieferant: Apollo Scientific
PC303488-1G PC303488-5G
APOSPC303488-1GEA 231 EUR
APOSPC303488-1G APOSPC303488-5G
3-Bromo-5,6-difluoro-2-methoxybenzyl alcohol
3-Bromo-5,6-difluoro-2-methoxybenzyl alcohol
Formel: C₈H₇BrF₂O₂
Molecular Weight: 253.041 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640082
CAS-Nummer: 1898381-62-8