3-Bromo-2,5,6-trifluorobenzamide
Lieferant: Apollo Scientific
PC303512-1G
PC303512-5G
APOSPC303512-1GEA
279
EUR
APOSPC303512-1G
APOSPC303512-5G
3-Bromo-2,5,6-trifluorobenzamide
3-Bromo-2,5,6-trifluorobenzamide
Formel:
C₇H₃BrF₃NO Molecular Weight: 254,004 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640104 CAS-Nummer: 1506289-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...