3-Bromo-2,5,6-trifluorobenzylamine

Lieferant: Apollo Scientific

PC303513-1G PC303513-5G
APOSPC303513-1GEA 418 EUR
APOSPC303513-1G APOSPC303513-5G
3-Bromo-2,5,6-trifluorobenzylamine
3-Bromo-2,5,6-trifluorobenzylamine
Formel: C₇H₅BrF₃N
Molecular Weight: 240,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30471770
CAS-Nummer: 1539083-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR