3-Bromo-4-(trifluoromethoxy)phenylacetic acid
Lieferant: Apollo Scientific
PC303526-1G
PC303526-5G
APOSPC303526-1GEA
418
EUR
APOSPC303526-1G
APOSPC303526-5G
3-Bromo-4-(trifluoromethoxy)phenylacetic acid
3-Bromo-4-(trifluoromethoxy)phenylacetic acid
Formel:
C₉H₆BrF₃O₃ Molecular Weight: 299,041 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640115 CAS-Nummer: 2169367-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...