5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: (5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)methanol

PC303541-1G PC303541-5G
APOSPC303541-1GEA 406 EUR
APOSPC303541-1G APOSPC303541-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅BrF₄O
Molecular Weight: 273,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640122
CAS-Nummer: 2090690-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR