5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetic acid

Lieferant: Apollo Scientific

PC303544-1G PC303544-5G
APOSPC303544-1GEA 430 EUR
APOSPC303544-1G APOSPC303544-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetic acid
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetic acid
Formel: C₉H₅BrF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD32640125
CAS-Nummer: 2384476-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR