3-Bromo-4-chloro-5-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific

PC303568-1G PC303568-5G
APOSPC303568-1GEA 418 EUR
APOSPC303568-1G APOSPC303568-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzylamine
3-Bromo-4-chloro-5-(trifluoromethyl)benzylamine
Formel: C₈H₆BrClF₃N
Molecular Weight: 288.492 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640146
CAS-Nummer: 2383229-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR