3-Bromo-4-chloro-5-(trifluoromethyl)benzonitrile 97%

Lieferant: Apollo Scientific

PC303572-1G PC303572-5G
APOSPC303572-1GEA 0 EUR
APOSPC303572-1G APOSPC303572-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzonitrile 97%
3-Bromo-4-chloro-5-(trifluoromethyl)benzonitrile
Formel: C₈H₂BrClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640150
CAS-Nummer: 2383568-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR