3-Bromo-2-chloro-5-(trifluoromethyl)benzamide
Lieferant: Apollo Scientific
PC303583-1G
PC303583-5G
APOSPC303583-1GEA
279
EUR
APOSPC303583-1G
APOSPC303583-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzamide
3-Bromo-2-chloro-5-(trifluoromethyl)benzamide
Formel:
C₈H₄BrClF₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640160 CAS-Nummer: 1344087-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...