3-Bromo-2-chloro-5-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC303585-1G PC303585-5G
APOSPC303585-1GEA 372 EUR
APOSPC303585-1G APOSPC303585-5G
3-Bromo-2-chloro-5-(trifluoromethyl)aniline
3-Bromo-2-chloro-5-(trifluoromethyl)aniline
Formel: C₇H₄BrClF₃N
Molecular Weight: 274,466 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28734627
CAS-Nummer: 1805577-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR