5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303589-1G PC303589-5G
APOSPC303589-1GEA 395 EUR
APOSPC303589-1G APOSPC303589-5G
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetonitrile 97%
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetonitrile
Formel: C₁₀H₇BrF₃N
Molecular Weight: 278.068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640164
CAS-Nummer: 2384383-24-6
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR