5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid 98%
Lieferant: Apollo Scientific
PC303590-1G
PC303590-5G
APOSPC303590-1GEA
364
EUR
APOSPC303590-1G
APOSPC303590-5G
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid 98%
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid
Formel:
C₁₀H₈BrF₃O₂ Molecular Weight: 297.069 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640165 CAS-Nummer: 2383520-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...