5-Bromo-2-methyl-3-(trifluoromethyl)benzylamine
Lieferant: Apollo Scientific
PC303591-1G
PC303591-5G
APOSPC303591-1GEA
430
EUR
APOSPC303591-1G
APOSPC303591-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzylamine
5-Bromo-2-methyl-3-(trifluoromethyl)benzylamine
Formel:
C₉H₉BrF₃N Molecular Weight: 268.074 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640166 CAS-Nummer: 2385458-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...