3-Bromo-4-methyl-5-(trifluoromethyl)benzamide
Lieferant: Apollo Scientific
PC303607-1G
PC303607-5G
APOSPC303607-1GEA
348
EUR
APOSPC303607-1G
APOSPC303607-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzamide
3-Bromo-4-methyl-5-(trifluoromethyl)benzamide
Formel:
C₉H₇BrF₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640180 CAS-Nummer: 2386432-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...