1H,1H,2'H,3H-Decafluorodipropyl ether 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1,1,2,3,3,6,6,7,7-Decafluoro-4-oxaheptane
PC3038-25G
PC3038-5G
APOSPC3038-25GEA
538
EUR
APOSPC3038-25G
APOSPC3038-5G
1H,1H,2'H,3H-Decafluorodipropyl ether 97%
1H,1H,2'H,3H-Decafluorodipropyl ether
Formel:
C₆H₄F₁₀O Molecular Weight: 282,079 g/mol Siedepunkt: 105,9 °C Dichte: 1,57 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00476471 CAS-Nummer: 65064-78-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...