5-Bromo-6-fluoronicotinic acid 98%
Lieferant: Apollo Scientific
PC305016-250MG
PC305016-1G
APOSPC305016-250MGEA
111
EUR
APOSPC305016-250MG
APOSPC305016-1G
5-Bromo-6-fluoronicotinic acid 98%
5-Bromo-6-fluoronicotinic acid
Formel:
C₆H₃BrFNO₂ Molecular Weight: 219,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11036154 CAS-Nummer: 29241-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...