2'-Amino-4'-(trifluoromethyl)acetanilide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[2-Amino-4-(trifluoromethyl)phenyl]acetamide

PC31160-100MG PC31160-1G PC31160-250MG
APOSPC31160-100MGEA 53.8 EUR
APOSPC31160-100MG APOSPC31160-1G APOSPC31160-250MG
2'-Amino-4'-(trifluoromethyl)acetanilide
2'-Amino-4'-(trifluoromethyl)acetanilide
Formel: C₉H₉F₃N₂O
Molecular Weight: 218.176 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00018052
CAS-Nummer: 97051-69-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR