5-Methyl-2-(trifluoromethyl)fur-3-yl isocyanate 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Isocyanato-5-methyl-2-(trifluoromethyl)furan
PC31425-1G
PC31425-250MG
PC31425-5G
APOSPC31425-1GEA
372
EUR
APOSPC31425-1G
APOSPC31425-250MG
APOSPC31425-5G
5-Methyl-2-(trifluoromethyl)fur-3-yl isocyanate 90%
5-Methyl-2-(trifluoromethyl)fur-3-yl isocyanate
May form small amount of sediment
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191.107 g/mol Siedepunkt: 60 °C (10 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD02677717 CAS-Nummer: 306935-03-5 UN: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...