6-Fluoro-4H-1,3-benzodioxine-8-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

PC31430-1G PC31430-5G PC31430-10G
APOSPC31430-1GEA 58 EUR
APOSPC31430-1G APOSPC31430-5G APOSPC31430-10G
6-Fluoro-4H-1,3-benzodioxine-8-carboxaldehyde
6-Fluoro-4H-1,3-benzodioxine-8-carboxaldehyde
Formel: C₉H₇FO₃
Molecular Weight: 182,148 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677682
CAS-Nummer: 306934-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR