6-Fluoro-8-(hydroxymethyl)-4H-1,3-benzodioxine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Fluoro-4H-1,3-benzodioxin-8-yl)methanol
PC31432-1G
PC31432-250MG
APOSPC31432-1GEA
132
EUR
APOSPC31432-1G
APOSPC31432-250MG
6-Fluoro-8-(hydroxymethyl)-4H-1,3-benzodioxine
6-Fluoro-8-(hydroxymethyl)-4H-1,3-benzodioxine
Formel:
C₉H₉FO₃ Molecular Weight: 184,164 g/mol Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677744 CAS-Nummer: 306934-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...