N-Benzyl-2,4,6-triphenylpyridinium tetrafluoroborate

Lieferant: Apollo Scientific
Achtung

PC31766-1G PC31766-250MG
APOSPC31766-1GEA 256 EUR
APOSPC31766-1G APOSPC31766-250MG
N-Benzyl-2,4,6-triphenylpyridinium tetrafluoroborate
N-Benzyl-2,4,6-triphenylpyridinium tetrafluoroborate
Formel: C₃₀H₂₄N·BF₄
Molecular Weight: 485,323 g/mol
Schmelzpunkt: 156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00960867
CAS-Nummer: 66310-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR