2-[(2-Aminoethyl)amino]-3-chloro-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine, 6-[(2-Aminoethyl)amino]-5-chloro-α,α,α-trifluoro-3-picoline

PC31984-1G PC31984-5G
APOSPC31984-1GEA 149 EUR
APOSPC31984-1G APOSPC31984-5G
2-[(2-Aminoethyl)amino]-3-chloro-5-(trifluoromethyl)pyridine
2-[(2-Aminoethyl)amino]-3-chloro-5-(trifluoromethyl)pyridine
Formel: C₈H₉ClF₃N₃
Molecular Weight: 239,625 g/mol
Siedepunkt: 96 °C (0,3 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00099696
CAS-Nummer: 219478-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR