4-(Difluoromethyl)sulphonylaniline

Lieferant: Apollo Scientific

PC32171-1G PC32171-5G
APOSPC32171-1GEA 176 EUR
APOSPC32171-1G APOSPC32171-5G
4-(Difluoromethyl)sulphonylaniline
4-(Difluoromethyl)sulphonylaniline
Formel: C₇H₇F₂NO₂S
Schmelzpunkt: 105…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00182675
CAS-Nummer: 24906-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR