Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Methoxycarbonyl)-4-phenyl-5-(trifluoromethyl)thiophene, Methyl 4-phenyl-5-(trifluoromethyl)-2-thenoate
PC32331-1G
PC32331-250MG
PC32331-5G
APOSPC32331-1GEA
98.9
EUR
APOSPC32331-1G
APOSPC32331-250MG
APOSPC32331-5G
Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate 95%
Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Formel:
C₁₃H₉F₃O₂S Molecular Weight: 286,27 g/mol Siedepunkt: 120…122 °C (2 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD00110251 CAS-Nummer: 237385-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...