Sodium tetrakis(4-fluorophenyl)borate hydrate 99+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrakis(4-fluorophenyl)boron sodium hydrate

PC3241-1G PC3241-5G
APOSPC3241-1GEA 243 EUR
APOSPC3241-1G APOSPC3241-5G
Sodium tetrakis(4-fluorophenyl)borate hydrate 99+%
Sodium tetrakis(4-fluorophenyl)borate hydrate

Nonionic surfactant

Formel: C₂₄H₁₆BF₄·Na·xH₂O
Molecular Weight: 414,19 g/mol (anhy)
Lagertemperatur: Kühlschrank
MDL: MFCD02183482
CAS-Nummer: 385812-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR