Ethyl perfluoroheptanoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl tridecafluoroheptanoate

PC3251-100G PC3251-250G
APOSPC3251-100GEA 248 EUR
APOSPC3251-100G APOSPC3251-250G
Ethyl perfluoroheptanoate 98%
Ethyl perfluoroheptanoate
Formel: C₉H₅F₁₃O₂
Molecular Weight: 392,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039242
CAS-Nummer: 41430-70-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR