3-(Chlorodifluoroacetyl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloro-2,2-difluoro-1-(imidazo[1,2-a]pyridin-3-yl)ethan-1-one

PC32713-1G PC32713-5G
APOSPC32713-1GEA 79.9 EUR
APOSPC32713-1G APOSPC32713-5G
3-(Chlorodifluoroacetyl)imidazo[1,2-a]pyridine
3-(Chlorodifluoroacetyl)imidazo[1,2-a]pyridine
Formel: C₉H₅ClF₂N₂O
Molecular Weight: 230.599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035867
CAS-Nummer: 219296-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR