2-Fluoro-6-iodobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Bromomethyl)-1-fluoro-3-iodobenzene, α-Bromo-2-fluoro-6-iodotoluene
PC32721-1G
PC32721-5G
PC32721-25G
APOSPC32721-1GEA
78.3
EUR
APOSPC32721-1G
APOSPC32721-5G
APOSPC32721-25G
2-Fluoro-6-iodobenzyl bromide
2-Fluoro-6-iodobenzyl bromide
Formel:
C₇H₅BrFI Molecular Weight: 314,921 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18396957 CAS-Nummer: 1231747-49-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...