4-Nitro-2-(trifluoromethyl)benzene-1,3-diamine 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Diamino-3-nitrobenzotrifluoride, 4-Nitro-2-(trifluoromethyl)phenylene-1,3-diamine
PC32722-1G
PC32722-250MG
APOSPC32722-1GEA
328
EUR
APOSPC32722-1G
APOSPC32722-250MG
4-Nitro-2-(trifluoromethyl)benzene-1,3-diamine 96%
4-Nitro-2-(trifluoromethyl)benzene-1,3-diamine
Formel:
C₇H₆F₃N₃O₂ Molecular Weight: 221,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25976386 CAS-Nummer: 1448858-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...