2-Amino-6-iodo-4-[(trifluoromethyl)thio]phenol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-3-iodo-5-[(trifluoromethyl)thio]aniline, 2-Amino-6-iodo-4-[(trifluoromethyl)sulphanyl]phenol

PC32724-1G PC32724-250MG
APOSPC32724-1GEA 859 EUR
APOSPC32724-1G APOSPC32724-250MG
2-Amino-6-iodo-4-[(trifluoromethyl)thio]phenol 95%
2-Amino-6-iodo-4-[(trifluoromethyl)thio]phenol
Formel: C₇H₅F₃INOS
Molecular Weight: 335,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976388
CAS-Nummer: 1448858-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR