2-Ethynyl-5-fluorobenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-2-formylphenylacetylene

PC32735-1G PC32735-250MG PC32735-5G
APOSPC32735-1GEA 290 EUR
APOSPC32735-1G APOSPC32735-250MG APOSPC32735-5G
2-Ethynyl-5-fluorobenzaldehyde 97%
2-Ethynyl-5-fluorobenzaldehyde
Formel: C₉H₅FO
Molecular Weight: 148.134 g/mol
Schmelzpunkt: 109 °C
Lagertemperatur: Kühlschrank
MDL: MFCD24674725
CAS-Nummer: 1015731-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR