3-Fluoro-5-iodobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Bromo-3-fluoro-5-iodotoluene, 1-(Bromomethyl)-3-fluoro-5-iodobenzene
PC32739-1G
PC32739-250MG
PC32739-5G
APOSPC32739-1GEA
223
EUR
APOSPC32739-1G
APOSPC32739-250MG
APOSPC32739-5G
3-Fluoro-5-iodobenzyl bromide 95%
3-Fluoro-5-iodobenzyl bromide
Formel:
C₇H₅BrFI Molecular Weight: 314,921 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18396960 CAS-Nummer: 1261759-75-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...