2-Iodo-4-(trifluoromethyl)benzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Iodo-4-(trifluoromethyl)phenyl)methanol, [2-Iodo-4-(trifluoromethyl)phenyl]methanol, 4-(Hydroxymethyl)-3-iodobenzotrifluoride
PC32741-1G
PC32741-250MG
PC32741-5G
APOSPC32741-1GEA
88.6
EUR
APOSPC32741-1G
APOSPC32741-250MG
APOSPC32741-5G
2-Iodo-4-(trifluoromethyl)benzyl alcohol
2-Iodo-4-(trifluoromethyl)benzyl alcohol
Formel:
C₈H₆F₃IO Molecular Weight: 302,032 g/mol Schmelzpunkt: 69…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398172 CAS-Nummer: 873005-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...