1-Bromo-2-methyl-4-(heptafluoropropan-2-yl)benzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-2-methylbenzene
PC32769-1G
PC32769-250MG
APOSPC32769-1GEA
719
EUR
APOSPC32769-1G
APOSPC32769-250MG
1-Bromo-2-methyl-4-(heptafluoropropan-2-yl)benzene 95%
1-Bromo-2-methyl-4-(heptafluoropropan-2-yl)benzene
Formel:
C₁₀H₆BrF₇ Molecular Weight: 339.047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29066216 CAS-Nummer: 353272-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...