2-Fluoro-5-cyclopropylphenol

Lieferant: Apollo Scientific
Achtung

PC32772-1G PC32772-250MG
APOSPC32772-1GEA 49.3 EUR
APOSPC32772-1G APOSPC32772-250MG
2-Fluoro-5-cyclopropylphenol
2-Fluoro-5-cyclopropylphenol
Formel: C₉H₉FO
Molecular Weight: 152,166 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD31382446
CAS-Nummer: 2379918-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR