2-Fluoro-6-cyclopropylphenol

Lieferant: Apollo Scientific
Achtung

PC32773-1G PC32773-250MG
APOSPC32773-1GEA 1230 EUR
APOSPC32773-1G APOSPC32773-250MG
2-Fluoro-6-cyclopropylphenol
2-Fluoro-6-cyclopropylphenol
Formel: C₉H₉FO
Molecular Weight: 152,166 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD24727590
CAS-Nummer: 911817-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR