7-Bromo-5-fluorobenzofuran-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC32792-1G
PC32792-250MG
APOSPC32792-1GEA
208
EUR
APOSPC32792-1G
APOSPC32792-250MG
7-Bromo-5-fluorobenzofuran-2-carboxylic acid
7-Bromo-5-fluorobenzofuran-2-carboxylic acid
Formel:
C₉H₄BrFO₃ Molecular Weight: 259,029 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD13187183 CAS-Nummer: 550998-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...