4’-Bromo-3’-chloro-2’-fluoroacetophenone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-3-chloro-2-fluorophenyl)ethanone

PC32796-1G PC32796-250MG PC32796-5G
APOSPC32796-1GEA 58 EUR
APOSPC32796-1G APOSPC32796-250MG APOSPC32796-5G
4’-Bromo-3’-chloro-2’-fluoroacetophenone 95%
4’-Bromo-3’-chloro-2’-fluoroacetophenone
Formel: C₈H₅BrClFO
Molecular Weight: 251.48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31922499
CAS-Nummer: 1690794-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR