5-Fluoro-2,3-dihydro-1,4-benzodioxine

Lieferant: Apollo Scientific
Achtung

PC32849-1G PC32849-250MG PC32849-5G
APOSPC32849-1GEA 91.5 EUR
APOSPC32849-1G APOSPC32849-250MG APOSPC32849-5G
5-Fluoro-2,3-dihydro-1,4-benzodioxine
5-Fluoro-2,3-dihydro-1,4-benzodioxine
Formel: C₈H₇FO₂
Molecular Weight: 154.138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27927511
CAS-Nummer: 69464-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR