3-(Dimethylamino)-1-(3-fluorophenyl)prop-2-en-1-one
Lieferant: Apollo Scientific
PC32872-1G
PC32872-250MG
PC32872-500MG
APOSPC32872-1GEA
771
EUR
APOSPC32872-1G
APOSPC32872-250MG
APOSPC32872-500MG
3-(Dimethylamino)-1-(3-fluorophenyl)prop-2-en-1-one
3-(Dimethylamino)-1-(3-fluorophenyl)prop-2-en-1-one
Formel:
C₁₁H₁₂FNO Molecular Weight: 193,217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570077 CAS-Nummer: 96604-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...