Ethyl 7-fluorobenzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC32890-1G PC32890-250MG
APOSPC32890-1GEA 116 EUR
APOSPC32890-1G APOSPC32890-250MG
Ethyl 7-fluorobenzofuran-2-carboxylate
Ethyl 7-fluorobenzofuran-2-carboxylate
Formel: C₁₁H₉FO₃
Molecular Weight: 208,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25956567
CAS-Nummer: 1207537-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR